2-Bromo-5-chloro-3-methylbenzo[b]thiophene ≥97%

Supplier: Matrix Scientific

084002-1G 084002-10G
102841-274EA 113.85 USD
102841-274 102841-276
2-Bromo-5-chloro-3-methylbenzo[b]thiophene ≥97%
2-Bromo-5-chloro-3-methylbenzo[b]thiophene

Hazard: Irritant

Formula: C₉H₆BrClS
MW: 261.57 g/mol
MDL Number: MFCD00126363
CAS Number: 175203-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR