4,5-Dibromo-2-phenylpyridazin-3(2H)-one ≥97%

Supplier: Matrix Scientific

083990-1G 083990-10G
102841-248EA 200 USD
102841-248 102841-250
4,5-Dibromo-2-phenylpyridazin-3(2H)-one ≥97%
4,5-Dibromo-2-phenylpyridazin-3(2H)-one

Hazard: Irritant

Formula: C₁₀H₆Br₂N₂O
MDL Number: MFCD00052907
CAS Number: 14305-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR