5-Bromouracil ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromopyrimidine-2,4(1H,3H)-dione, Bromouracil

083985-100G 083985-500G
102841-232EA 93.85 USD
102841-232 102841-234
5-Bromouracil ≥97%
5-Bromouracil

Hazard: Irritant

Formula: C₄H₃BrN₂O₂
MW: 190.98 g/mol
MDL Number: MFCD00006017
CAS Number: 51-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR