2-Chloro-5-propylpyrimidine ≥97%

Supplier: Matrix Scientific

083969-1G 083969-10G
102841-196EA 92.31 USD
102841-196 102841-198
2-Chloro-5-propylpyrimidine ≥97%
2-Chloro-5-propylpyrimidine

Hazard: Irritant

Formula: C₇H₉ClN₂
MW: 156.61
MDL Number: MFCD00799403
CAS Number: 219555-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR