4,5-Diiodo-2-methylimidazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4,5-Diiodo-2-methyl-1H-imidazole

083941-1G 083941-10G
102841-132EA 2120 USD
102841-132 102841-134
4,5-Diiodo-2-methylimidazole ≥97%
4,5-Diiodo-2-methylimidazole

Hazard: Irritant

Formula: C₄H₄I₂N₂
MW: 333.9 g/mol
MDL Number: MFCD00464337
CAS Number: 73746-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR