tert-Butyl-4-bromo-1H-pyrazole-1-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-bromo-1H-pyrazole-1-carboxylate, 4-Bromo-1H-pyrazole, N1-BOC protected

083921-1G 083921-10G
102841-080EA 156.92 USD
102841-080 102841-082
tert-Butyl-4-bromo-1H-pyrazole-1-carboxylate ≥97%
tert-Butyl-4-bromo-1H-pyrazole-1-carboxylate

Hazard: Irritant

Formula: C₈H₁₁BrN₂O₂
MDL Number: MFCD09999163
CAS Number: 1150271-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR