5-Chloro-1,3-dimethyl-1H-pyrazole ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-1,3-dimethylpyrazole

083918-1G 083918-10G 083918-50G
102841-070EA 101.54 USD
102841-070 102841-072 102841-074
5-Chloro-1,3-dimethyl-1H-pyrazole ≥97%
5-Chloro-1,3-dimethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₇ClN₂
MW: 130.58 g/mol
MDL Number: MFCD00051650
CAS Number: 54454-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR