4-Bromo-2-(trichloroacetyl)pyrrole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(4-Bromo-1H-pyrrol-2-yl)-2,2,2-trichloroethanone

083910-1G 083910-10G
102841-048EA 467.69 USD
102841-048 102841-050
4-Bromo-2-(trichloroacetyl)pyrrole ≥97%
4-Bromo-2-(trichloroacetyl)pyrrole

Hazard: Irritant

Formula: C₆H₃BrCl₃NO
MW: 291.3571 g/mol
MDL Number: MFCD00832836
CAS Number: 72652-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR