2-(5-Bromothien-2-yl)pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(5-Bromothiophen-2-yl)pyridine

083904-1G 083904-10G
102841-032EA 156.92 USD
102841-032 102841-034
2-(5-Bromothien-2-yl)pyridine ≥97%
2-(5-Bromothien-2-yl)pyridine

Hazard: Irritant

Formula: C₉H₆BrNS
MW: 240.13 g/mol
MDL Number: MFCD00120466
CAS Number: 123784-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR