3,4-Dichloropyridine hydrochloride ≥98%

Supplier: Matrix Scientific

083890-1G 083890-10G 083890-50G
102840-992EA 200 USD
102840-992 102840-994 102840-996
3,4-Dichloropyridine hydrochloride ≥98%
3,4-Dichloropyridine hydrochloride

Hazard: Irritant

Formula: C₅H₄Cl₃N
MDL Number: MFCD06656018
CAS Number: 159732-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR