2-Iodopyridine ≥97%

Supplier: Matrix Scientific

083888-10G 083888-50G
102840-986EA 516.92 USD
102840-986 102840-988
2-Iodopyridine ≥97%
2-Iodopyridine

Hazard: Irritant

Formula: C₅H₄IN
MW: 205 g/mol
MDL Number: MFCD00464928
CAS Number: 5029-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR