5-Chloro-2-hydroxypyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-6-pyridinol, 5-Chloro-2-pyridinol, 3-Chloro-6-hydroxypyridine

083875-1G 083875-10G 083875-50G
102840-952EA 15.38 USD
102840-952 102840-954 102840-956
5-Chloro-2-hydroxypyridine ≥98%
5-Chloro-2-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄ClNO
MW: 129.55 g/mol
MDL Number: MFCD00006275
CAS Number: 4214-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR