3-Chloro-2-hydroxypyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2-pyridinol

083858-1G 083858-10G
102840-914EA 127.69 USD
102840-914 102840-916
3-Chloro-2-hydroxypyridine ≥97%
3-Chloro-2-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄ClNO
MW: 129.55 g/mol
MDL Number: MFCD01646109
CAS Number: 13466-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR