2-Bromo-4-nitropyridine ≥97%

Supplier: Matrix Scientific

083831-1G 083831-10G 083831-50G
102840-850EA 143.08 USD
102840-850 102840-852 102840-854
2-Bromo-4-nitropyridine ≥97%
2-Bromo-4-nitropyridine

Hazard: Irritant

Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD03701383
CAS Number: 6945-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR