2,3,5,6-Tetrachloropyridine ≥97%

Supplier: Matrix Scientific

083829-1G 083829-10G 083829-50G
102840-842EA 49.23 USD
102840-842 102840-844 102840-846
2,3,5,6-Tetrachloropyridine ≥97%
2,3,5,6-Tetrachloropyridine

Hazard: Irritant

Formula: C₅HCl₄N
MW: 216.88 g/mol
MDL Number: MFCD00092428
CAS Number: 2402-79-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR