5-tert-Butyl-2-iodo-1,3-dimethylbenzene ≥97%

Supplier: Matrix Scientific

083824-1G 083824-50G
102840-828EA 156.92 USD
102840-828 102840-832
5-tert-Butyl-2-iodo-1,3-dimethylbenzene ≥97%
5-tert-Butyl-2-iodo-1,3-dimethylbenzene

Hazard: Irritant

Formula: C₁₂H₁₇I
MW: 288.17 g/mol
MDL Number: MFCD00052173
CAS Number: 5122-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR