5-Butyl-2-iodotoluene ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Butyl-1-iodo-2-methylbenzene
083812-1G
083812-10G
102840-802EA
48.38
USD
102840-802
102840-804
5-Butyl-2-iodotoluene ≥97%
5-Butyl-2-iodotoluene
Hazard: Irritant
Formula:
C₁₁H₁₅I MW: 274.14 g/mol |
CAS Number:
175278-38-3 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...