Tris(4-chlorophenyl)phosphine ≥98%

Supplier: Matrix Scientific

Synonyms: Tri(p-chlorophenyl)phosphine, Tris(p-chlorophenyl)phosphine

083808-1G 083808-10G
102840-786EA 113.85 USD
102840-786 102840-788
Tris(4-chlorophenyl)phosphine ≥98%
Tris(4-chlorophenyl)phosphine

Hazard: Irritant

Formula: C₁₈H₁₂Cl₃P
MW: 365.63 g/mol
MDL Number: MFCD00013639
CAS Number: 1159-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR