1-Bromo-2-methyl-4-(trifluoromethoxy)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-(trifluoromethoxy)toluene

083806-1G 083806-10G
102840-780EA 29.23 USD
102840-780 102840-782
1-Bromo-2-methyl-4-(trifluoromethoxy)benzene ≥97%
1-Bromo-2-methyl-4-(trifluoromethoxy)benzene

Hazard: Irritant-Harmful/Light Sensitive

Formula: C₈H₆BrF₃O
MW: 255.0319 g/mol
Density: 1.559
Flash Pt: 78.3 °C
MDL Number: MFCD01631545
CAS Number: 261951-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR