4-(3-Bromophenyl)morpholine ≥97%

Supplier: Matrix Scientific

083801-1G 083801-10G
102840-768EA 56.92 USD
102840-768 102840-770
4-(3-Bromophenyl)morpholine ≥97%
4-(3-Bromophenyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MW: 242.1100 g/mol
MDL Number: MFCD04112585
CAS Number: 197846-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR