2,5-Dichloro-p-xylene ≥97%

Supplier: Matrix Scientific

083792-10G 083792-50G
102840-742EA 78.46 USD
102840-742 102840-744
2,5-Dichloro-p-xylene ≥97%
2,5-Dichloro-p-xylene

Hazard: Irritant

Formula: C₈H₈Cl₂
MDL Number: MFCD00000610
CAS Number: 1124-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR