5-(2-Chlorophenyl)oxazole ≥97%

Supplier: Matrix Scientific

083782-1G 083782-10G
102840-712EA 255.38 USD
102840-712 102840-714
5-(2-Chlorophenyl)oxazole ≥97%
5-(2-Chlorophenyl)oxazole

Hazard: Irritant

Formula: C₉H₆ClNO
MW: 179.6030 g/mol
MDL Number: MFCD00084973
CAS Number: 89808-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR