3-tert-Butyl-1-bromobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-tert-butylbenzene, 1-Bromo-3-tert-butylbenzene, 1-Bromo-3-(tert-butyl)benzene, 2-(3-Bromophenyl)-2-methylpropane

083778-1G 083778-10G 083778-50G
102840-698EA 44.62 USD
102840-698 102840-700 102840-702
3-tert-Butyl-1-bromobenzene ≥97%
3-tert-Butyl-1-bromobenzene

Hazard: Irritant

Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
MDL Number: MFCD00234661
CAS Number: 3972-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR