2-(4-Bromophenoxy)-4-(trifluoromethyl)pyrimidine ≥97%

Supplier: Matrix Scientific

083776-1G 083776-10G
102840-692EA 383.08 USD
102840-692 102840-694
2-(4-Bromophenoxy)-4-(trifluoromethyl)pyrimidine ≥97%
2-(4-Bromophenoxy)-4-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₁H₆BrF₃N₂O
MDL Number: MFCD08235249
CAS Number: 914636-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR