4-Bromophenylsulfur pentafluoride ≥99%

Supplier: Matrix Scientific

083770-1G 083770-5G
102840-678EA 312.31 USD
102840-678 102840-680
4-Bromophenylsulfur pentafluoride ≥99%
4-Bromophenylsulphur pentafluoride

Hazard: Toxic/Refrigerate

Formula: C₆H₄BrF₅S
Boiling Pt: 77-78°C/10mm
Flash Pt: 84°C
MDL Number: MFCD03425928
CAS Number: 774-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR