3-Ethyl-2-iodotoluene ≥97%

Supplier: Matrix Scientific

083764-1G 083764-10G 083764-50G
102840-660EA 86.15 USD
102840-660 102840-662 102840-664
3-Ethyl-2-iodotoluene ≥97%
3-Ethyl-2-iodotoluene

Hazard: Irritant

Formula: C₉H₁₁I
MDL Number: MFCD00052855
CAS Number: 175277-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR