3,5-Dichloro-2-iodotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 1,3-Dichloro-4-iodo-5-methylbenzene

083754-1G 083754-10G
102840-636EA 127.69 USD
102840-636 102840-638
3,5-Dichloro-2-iodotoluene ≥97%
3,5-Dichloro-2-iodotoluene

Hazard: Irritant

Formula: C₇H₅Cl₂I
MW: 286.93 g/mol
MDL Number: MFCD00052856
CAS Number: 175277-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR