2-Chloro-5-iodobenzotrifluoride ≥97%
Supplier: Matrix Scientific
083752-1G
083752-10G
102840-630EA
127.69
USD
102840-630
102840-632
2-Chloro-5-iodobenzotrifluoride ≥97%
2-Chloro-5-iodobenzotrifluoride
Hazard: Irritant
Formula:
C₇H₃ClF₃I |
MDL Number:
MFCD09037801 CAS Number: 260355-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...