3-Bromo-5-methylbenzotrifluoride ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Bromo-3-methyl-5-(trifluoromethyl)benzene
083747-1G
083747-10G
102840-618EA
241.54
USD
102840-618
102840-620
3-Bromo-5-methylbenzotrifluoride ≥97%
3-Bromo-5-methylbenzotrifluoride
Hazard: Irritant
Formula:
C₈H₆BrF₃ MW: 239.04 g/mol |
MDL Number:
MFCD01631585 CAS Number: 86845-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...