1-Ethyl-4-(2-oxo-1,2-diphenylethyl)succinate ≥97%

Supplier: Matrix Scientific

083738-1G 083738-10G
102840-594EA 467.69 USD
102840-594 102840-596
1-Ethyl-4-(2-oxo-1,2-diphenylethyl)succinate ≥97%
1-Ethyl-4-(2-oxo-1,2-diphenylethyl)succinate

Hazard: Irritant

Formula: C₂₀H₂₀O₅
MW: 340.38 g/mol
MDL Number: MFCD04112426
CAS Number: 499785-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR