Isopropyl DL-leucate ≥97%

Supplier: Matrix Scientific

Synonyms: Isopropyl DL-2-Hydroxy-4-methylvalerate, DL-Leucic Acid Isopropyl Ester

083730-1G 083730-10G
102840-568EA 72.31 USD
102840-568 102840-570
Isopropyl DL-leucate ≥97%
Isopropyl DL-leucate

Hazard: Irritant

Formula: C₉H₁₈O₃
MW: 174.24 g/mol
MDL Number: MFCD00059361
CAS Number: 156276-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR