tert-Butyl aminosulfonylcarbamate ≥97%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl sulphamoylcarbamate
083720-1G
083720-5G
102840-534EA
63.08
USD
102840-534
102840-536
tert-Butyl aminosulfonylcarbamate ≥97%
tert-Butyl aminosulfonylcarbamate
Hazard: Irritant
Formula:
C₅H₁₂N₂O₄S MW: 196.23 g/mol |
MDL Number:
MFCD11616268 CAS Number: 148017-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...