Diethyl cyclopentylmalonate ≥97%

Supplier: Matrix Scientific

083717-1G 083717-10G
102840-526EA 78.46 USD
102840-526 102840-528
Diethyl cyclopentylmalonate ≥97%
Diethyl cyclopentylmalonate

Hazard: Irritant

Formula: C₁₂H₂₀O₄
Density: 1.03
MDL Number: MFCD00059514
CAS Number: 18928-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR