Methyl-5-methyl-2-hexenoate ≥97%
Supplier: Matrix Scientific
083704-1G
083704-10G
083704-50G
102840-476EA
233.85
USD
102840-476
102840-478
102840-480
Methyl-5-methyl-2-hexenoate ≥97%
Methyl-5-methyl-2-hexenoate
Hazard: Irritant
Formula:
C₈H₁₄O₂ |
MDL Number:
MFCD00137613 CAS Number: 75513-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...