Vinyl crotonate ≥97%

Supplier: Matrix Scientific

Synonyms: Crotonic Acid Vinyl Ester

083703-1G 083703-10G 083703-50G
102840-470EA 72.31 USD
102840-470 102840-472 102840-474
Vinyl crotonate ≥97%
Vinyl crotonate

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00009288
CAS Number: 14861-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR