Ethyl-2-amino-3-hydroxypropanoate hydrochloride ≥97%

Supplier: Matrix Scientific

083698-1G 083698-10G
102840-454EA 86.15 USD
102840-454 102840-456
Ethyl-2-amino-3-hydroxypropanoate hydrochloride ≥97%
Ethyl-2-amino-3-hydroxypropanoate hydrochloride

Hazard: Irritant

Formula: C₅H₁₁NO₃·HCl
MDL Number: MFCD00067544
CAS Number: 3940-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR