Dimethyl aminomalonate ≥97%

Supplier: Matrix Scientific

083697-1G 083697-10G
102840-450EA 426.15 USD
102840-450 102840-452
Dimethyl aminomalonate ≥97%
Dimethyl aminomalonate

Hazard: Irritant

Formula: C₅H₉NO₄
MW: 147.13 g/mol
MDL Number: MFCD00829425
CAS Number: 53704-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR