{[3-Ethoxy-2-(ethoxycarbonyl)-3-oxoprop-1-enyl]amino}acetic acid ≥97%

Supplier: Matrix Scientific

083691-1G 083691-10G
102840-428EA 113.85 USD
102840-428 102840-430
{[3-Ethoxy-2-(ethoxycarbonyl)-3-oxoprop-1-enyl]amino}acetic acid ≥97%
{[3-Ethoxy-2-(ethoxycarbonyl)-3-oxoprop-1-enyl]amino}acetic acid

Hazard: Irritant

Formula: C₁₀H₁₅NO₆
MDL Number: MFCD01909989
CAS Number: 54132-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR