Ethyl-2-phenylpropionate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-phenylpropanoate
083687-1G
083687-10G
102840-414EA
98.46
USD
102840-414
102840-416
Ethyl-2-phenylpropionate ≥97%
Ethyl-2-phenylpropionate
Hazard: Irritant
Formula:
C₁₁H₁₄O₂ MW: 178.2277 g/mol |
MDL Number:
MFCD00795471 CAS Number: 2510-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...