Diethyl isopropylidenemalonate ≥97%

Supplier: Matrix Scientific

083684-1G 083684-10G 083684-50G
102840-402EA 72.31 USD
102840-402 102840-404 102840-406
Diethyl isopropylidenemalonate ≥97%
Diethyl isopropylidenemalonate

Hazard: Irritant

Formula: C₁₀H₁₆O₄
MDL Number: MFCD00009147
CAS Number: 6802-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR