Methyl-6-hydroxy-2-naphthoate ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxy-2-naphthoic acid methyll ester, Methyl 6-hydroxy-2-naphthoate

083670-1G 083670-10G
102840-364EA 156.92 USD
102840-364 102840-366
Methyl-6-hydroxy-2-naphthoate ≥97%
Methyl 6-hydroxy-2-naphthoate

Hazard: Irritant

Formula: C₁₂H₁₀O₃
MW: 202.2060 g/mol
MDL Number: MFCD00100526
CAS Number: 17295-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR