Ethyl-3-(butylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate ≥97%

Supplier: Matrix Scientific

083652-1G 083652-10G
102840-326EA 113.85 USD
102840-326 102840-328
Ethyl-3-(butylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate ≥97%
Ethyl-3-(butylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₀O₃S₂
MDL Number: MFCD00085061
CAS Number: 172516-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR