Methyl-1-methylcyclopropane-1-carboxylate ≥97%
Supplier: Matrix Scientific
083618-1G
083618-10G
102840-240EA
156.92
USD
102840-240
102840-242
Methyl-1-methylcyclopropane-1-carboxylate ≥97%
Methyl-1-methylcyclopropane-1-carboxylate
Hazard: Irritant
Formula:
C₆H₁₀O₂ |
MDL Number:
MFCD00191549 CAS Number: 6206-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...