Ethyl-3-oxocyclohexanecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-oxocyclohexane-1-carboxylate

083606-1G 083606-5G
102840-202EA 176.92 USD
102840-202 102840-204
Ethyl-3-oxocyclohexanecarboxylate ≥97%
Ethyl-3-oxocyclohexanecarboxylate

Hazard: Irritant

Formula: C₉H₁₄O₃
MDL Number: MFCD00205583
CAS Number: 33668-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR