Ethyl morpholine-2-carboxylate ≥97%

Supplier: Matrix Scientific

083576-1G 083576-10G
102840-134EA 256.92 USD
102840-134 102840-136
Ethyl morpholine-2-carboxylate ≥97%
Ethyl morpholine-2-carboxylate

Hazard: Irritant

Formula: C₇H₁₃NO₃
MDL Number: MFCD08751336
CAS Number: 135782-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR