Ethyl piperazin-1-ylacetate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(piperazin-1-yl)acetate

083574-1G 083574-10G
102840-128EA 86.15 USD
102840-128 102840-130
Ethyl piperazin-1-ylacetate ≥97%
Ethyl piperazin-1-ylacetate

Hazard: Irritant

Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
MDL Number: MFCD00040739
CAS Number: 40004-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR