tert-Butyl-3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(2-Methoxy-2-oxoethyl)piperazine, N1-BOC protected

083568-1G 083568-10G
102840-110EA 590.77 USD
102840-110 102840-112
tert-Butyl-3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate ≥97%
tert-Butyl-3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₄
MW: 258.31 g/mol
MDL Number: MFCD01861213
CAS Number: 183742-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR