Methyl isoquinoline-3-carboxylate ≥97%

Supplier: Matrix Scientific

083547-1G 083547-10G
102840-056EA 84.62 USD
102840-056 102840-058
Methyl isoquinoline-3-carboxylate ≥97%
Methyl isoquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₂
MW: 187.2000 g/mol
MDL Number: MFCD00075138
CAS Number: 27104-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR