Methyl-2,1,3-benzothiadiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2,1,3-benzothiadiazole-5-carboxylate

083543-1G 083543-10G
102840-046EA 170.77 USD
102840-046 102840-048
Methyl-2,1,3-benzothiadiazole-5-carboxylate ≥97%
Methyl-2,1,3-benzothiadiazole-5-carboxylate

Hazard: Irritant

Formula: C₈H₆N₂O₂S
MW: 194.21 g/mol
MDL Number: MFCD00173948
CAS Number: 175204-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR