Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

083504-1G 083504-10G
102839-956EA 101.54 USD
102839-956 102839-958
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate ≥97%

Hazard: Irritant

Formula: C₇H₉N₃O₂S
MDL Number: MFCD00023227
CAS Number: 774-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR